Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(4-Hydroxyphenylazo)benzoic acid

Catalog No. H51265G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H51265G 5 g
H512625G 25 g
H51261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H51265G Supplier Sigma Aldrich Supplier No. H51265G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(4-Hydroxyphenylazo)benzoic acid

Specifications

CAS 1634-82-8
Melting Point 204°C to 208°C (lit.)
Molecular Formula C13H10N2O3
Linear Formula HOC6H4N=NC6H4CO2H
MDL Number MFCD00002428
Quantity 5 g
Synonym 2-(4 -Hydroxybenzeneazo)benzoic acid; 4 -Hydroxyazobenzene-2-carboxylic acid; HABA; HBABA
Molecular Weight (g/mol) 242.23
Percent Purity ≥95% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.