Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-Imidazolidinedione, 5-[[4-[4-hydroxy-3-(1-methylethyl)phenoxy]-3,5-diiodophenyl] | 1370363-74-8 | 98.0% | 50 MG
SDP

Catalog No. 5000338619
Encompass_Preferred

CO23 is a selective thyroid hormone receptor (TR) α agonist used for growth and development regulation. In vivo studies in hypothyroid Wistar rats showed that CO23 administered intraperitoneally achieved approximately 50% cholesterol reduction at doses of 0.8 nmol/g and higher, and increased Dio1 mRNA levels significantly.

  • Selective thyroid hormone receptor (TR) α agonist
  • Regulates growth and development
  • Achieves approximately 50% cholesterol reduction in vivo
  • Increases Dio1 mRNA levels in vivo
  • Purity: 98.0%
  • CAS number: 1370363-74-8

Catalog No. 50-003-38619 Supplier Medchemexpress LLC Supplier No. HY13001250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CO23 is a selective thyroid hormone receptor (TR) α agonist used for growth and development regulation. In vivo studies in hypothyroid Wistar rats showed that CO23 administered intraperitoneally achieved approximately 50% cholesterol reduction at doses of 0.8 nmol/g and higher, and increased Dio1 mRNA levels significantly.

  • Selective thyroid hormone receptor (TR) α agonist
  • Regulates growth and development
  • Achieves approximately 50% cholesterol reduction in vivo
  • Increases Dio1 mRNA levels in vivo
  • Purity: 98.0%
  • CAS number: 1370363-74-8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.