Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-imidazolidinedione, 5-phenyl- | 89-24-7 | MFCD00037881 | ≥97.0% | 176.18 g/mol | C9H8N2O2 | 25MG
SDP

Catalog No. 5000239071
Encompass_Preferred

5-Phenylhydantoin is an antiepileptic research compound and a metabolite of mephenytoin, supplied as a white to off-white solid for research and analytical use only. It has chemical formula C9H8N2O2, molecular weight approximately 176.18 g/mol, CAS 89-24-7, and a reported purity of ≥97% (NMR). Recommended storage: powder at -20°C for long-term stability or 4°C for shorter-term storage; in solvent store at -80°C for extended periods or -20°C for short-term use.

  • Antiepileptic compound and metabolite of mephenytoin.
  • Reported purity ≥97% by NMR.
  • White to off-white solid appearance.
  • Chemical formula C9H8N2O2 and molecular weight ~176.18 g/mol.
  • Recommended storage conditions for powder and solutions to maintain stability.

Catalog No. 50-002-39071 Supplier Medchemexpress LLC Supplier No. HY12578025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Phenylhydantoin is an antiepileptic research compound and a metabolite of mephenytoin, supplied as a white to off-white solid for research and analytical use only. It has chemical formula C9H8N2O2, molecular weight approximately 176.18 g/mol, CAS 89-24-7, and a reported purity of ≥97% (NMR). Recommended storage: powder at -20°C for long-term stability or 4°C for shorter-term storage; in solvent store at -80°C for extended periods or -20°C for short-term use.

  • Antiepileptic compound and metabolite of mephenytoin.
  • Reported purity ≥97% by NMR.
  • White to off-white solid appearance.
  • Chemical formula C9H8N2O2 and molecular weight ~176.18 g/mol.
  • Recommended storage conditions for powder and solutions to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.