Learn More
Medchemexpress LLC 2-(4-isothiocyanatobenzyl)-1,4,7-triazacyclononane-1,4,7-triacetic acid trihydrochloride | 1583289-85-3 | 99.6% | 559.89 | C20H29Cl3N4O6S | 10mg
SDP
Catalog No. 5000268449
Encompass_Preferred
p-SCN-Bn-NOTA trihydrochloride is a macrocyclic chelator p-SCN-Bn-NOTA trihydrochloride can be covalently coupled to molecules such as peptides through the thiocyanate group to form hexacoordinate copper (such as 64Cu) complexes p-SCN-Bn-NOTA trihydrochloride specifically binds to GRPR or EGFR highly expressed on the surface of tumor cells mediating tumor enrichment of radioactive probes p-SCN-Bn-NOTA trihydrochloride can be used to study malignant tumors expressing GRPR or EGFR such as prostate cancer and colorectal cancer[1][2]
Catalog No. 50-002-68449 Supplier Medchemexpress LLC Supplier No. HYW78208310MG
p-SCN-Bn-NOTA trihydrochloride is a macrocyclic chelator p-SCN-Bn-NOTA trihydrochloride can be covalently coupled to molecules such as peptides through the thiocyanate group to form hexacoordinate copper (such as 64Cu) complexes p-SCN-Bn-NOTA trihydrochloride specifically binds to GRPR or EGFR highly expressed on the surface of tumor cells mediating tumor enrichment of radioactive probes p-SCN-Bn-NOTA trihydrochloride can be used to study malignant tumors expressing GRPR or EGFR such as prostate cancer and colorectal cancer[1][2]