Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Lutidine, 98+%, Thermo Scientific™

Catalog No. AC125180010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC125180010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC125180010 Supplier Thermo Scientific Chemicals Supplier No. 125180010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-47-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006337
InChI Key JYYNAJVZFGKDEQ-UHFFFAOYSA-N
Synonym 2,4-lutidine, pyridine, 2,4-dimethyl, alpha,gamma-dimethylpyridine, 2,4-lutidene, unii-83903uj0ww, 2,4-dimethyl-pyridine, .alpha.,.gamma.-dimethylpyridine, 2,4-dimethyl pyridine, 2.4-lutidine, 2,4 lutidine
PubChem CID 7936
IUPAC Name 2,4-dimethylpyridine
SMILES CC1=CC=NC(C)=C1

Specifications

Melting Point -60.0°C
Density 0.9200g/mL
Boiling Point 159.0°C
Flash Point 47°C
Refractive Index 1.4990 to 1.5010
Quantity 1 L
Specific Gravity 0.92
Solubility Information Solubility in water: 15 g/100 g (20°C). Other solubilities: soluble in alcohol, ether, acetone and, benzene
Formula Weight 107.15
Chemical Name or Material 2,4-Lutidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.