Learn More
Medchemexpress LLC HY-W007686 500mg Medchemexpress, Imidazoleacetic acid (hydrochloride) CAS:3251-69-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW007686500MG
Encompass_Preferred
Medchemexpress, HY-W004206 100mg 2-(4-Methoxyphenyl)acetic acid CAS:104-01-8 2-(4-Methoxyphenyl)acetic acid is a plasma metabolite, with high sensitivity and specificity value as a biomarker for discriminating between NSCLC and healthy controls. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0651