Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-Methoxyphenyl)acetic acid | 104-01-8 | 166.17 | 25 G
SDP

Catalog No. 5000448214
Encompass_Preferred

2-(4-Methoxyphenyl)acetic acid is a plasma metabolite with high sensitivity and specificity, serving as a biomarker for distinguishing between non-small cell lung cancer (NSCLC) and healthy individuals. It may play a protective role in preventing lung cancer development. This compound is for research use only.

  • Biomarker for non-small cell lung cancer (NSCLC)
  • High sensitivity and specificity for discriminating NSCLC
  • May prevent lung cancer development
  • Molecular formula C9H10O3
  • Molecular weight 166.17
  • Appearance: solid, white to off-white
  • Store solid at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-48214 Supplier Medchemexpress LLC Supplier No. 25GHYW004206
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-(4-Methoxyphenyl)acetic acid is a plasma metabolite with high sensitivity and specificity, serving as a biomarker for distinguishing between non-small cell lung cancer (NSCLC) and healthy individuals. It may play a protective role in preventing lung cancer development. This compound is for research use only.

  • Biomarker for non-small cell lung cancer (NSCLC)
  • High sensitivity and specificity for discriminating NSCLC
  • May prevent lung cancer development
  • Molecular formula C9H10O3
  • Molecular weight 166.17
  • Appearance: solid, white to off-white
  • Store solid at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.