Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-Methoxyphenyl)acetic acid | 104-01-8 | 166.18 | 10 G
SDP

Catalog No. 5000449239
Encompass_Preferred

2-(4-Methoxyphenyl)acetic acid is a plasma metabolite with high sensitivity and specificity as a biomarker for distinguishing between non-small cell lung cancer (NSCLC) and healthy controls. It may also play a protective role in preventing the development of lung cancer.

  • Plasma metabolite
  • Biomarker for non-small cell lung cancer
  • May prevent lung cancer development
  • Solid appearance
  • White to off-white color
  • Melting point 84-86 °C
  • Boiling point 140 °C at 760 mmHg
  • Chemically stable under recommended storage conditions
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-49239 Supplier Medchemexpress LLC Supplier No. HYW00420610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(4-Methoxyphenyl)acetic acid is a plasma metabolite with high sensitivity and specificity as a biomarker for distinguishing between non-small cell lung cancer (NSCLC) and healthy controls. It may also play a protective role in preventing the development of lung cancer.

  • Plasma metabolite
  • Biomarker for non-small cell lung cancer
  • May prevent lung cancer development
  • Solid appearance
  • White to off-white color
  • Melting point 84-86 °C
  • Boiling point 140 °C at 760 mmHg
  • Chemically stable under recommended storage conditions
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.