Learn More
Medchemexpress LLC 2-(4-Methoxyphenyl)acetic acid | 104-01-8 | MFCD00004345 | 166.17 | 100 G
SDP
Catalog No. 5000447467
Encompass_Preferred
2-(4-Methoxyphenyl)acetic acid is a plasma metabolite that serves as a biomarker for distinguishing between Non-Small Cell Lung Cancer (NSCLC) and healthy individuals, showing high sensitivity and specificity. It may also have a protective role in preventing the development of lung cancer.
  • Biomarker for distinguishing between non-small cell lung cancer and healthy individuals.
  • Exhibits high sensitivity and specificity.
  • May play a protective role in preventing lung cancer development.
  • Appears as a white to off-white solid.
  • Classified as a ketone, aldehyde, and acid.
  • Acts as an endogenous human metabolite.
  • Shows anticancer activity.
  • Demonstrates antineuroinflammatory activity.
Catalog No. 50-004-47467 Supplier Medchemexpress LLC Supplier No. HYW004206100G
Add to Cart
Add to Cart
2-(4-Methoxyphenyl)acetic acid is a plasma metabolite that serves as a biomarker for distinguishing between Non-Small Cell Lung Cancer (NSCLC) and healthy individuals, showing high sensitivity and specificity. It may also have a protective role in preventing the development of lung cancer.
  • Biomarker for distinguishing between non-small cell lung cancer and healthy individuals.
  • Exhibits high sensitivity and specificity.
  • May play a protective role in preventing lung cancer development.
  • Appears as a white to off-white solid.
  • Classified as a ketone, aldehyde, and acid.
  • Acts as an endogenous human metabolite.
  • Shows anticancer activity.
  • Demonstrates antineuroinflammatory activity.