Learn More
Medchemexpress LLC 2-(4-Methoxyphenyl)acetic acid | 104-01-8 | MFCD00004345 | 166.18 | 50 G
SDP
Catalog No. 5000449106
Encompass_Preferred
2-(4-Methoxyphenyl)acetic acid is a plasma metabolite with high sensitivity and specificity as a biomarker for discriminating between non-small cell lung cancer (NSCLC) and healthy controls. It may play a protective role in preventing the development of lung cancer.
  • Plasma metabolite and biomarker for discriminating between NSCLC and healthy controls.
  • Shows anticancer activity against human A549, SK-MEL-2, SK-OV-3, and XF498 cells.
  • Exhibits antineuroinflammatory activity in mouse N9 cells.
  • May protect against the development of lung cancer.
Catalog No. 50-004-49106 Supplier Medchemexpress LLC Supplier No. HYW00420650G
Add to Cart
Add to Cart
2-(4-Methoxyphenyl)acetic acid is a plasma metabolite with high sensitivity and specificity as a biomarker for discriminating between non-small cell lung cancer (NSCLC) and healthy controls. It may play a protective role in preventing the development of lung cancer.
  • Plasma metabolite and biomarker for discriminating between NSCLC and healthy controls.
  • Shows anticancer activity against human A549, SK-MEL-2, SK-OV-3, and XF498 cells.
  • Exhibits antineuroinflammatory activity in mouse N9 cells.
  • May protect against the development of lung cancer.