Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methoxyphenyl)indole 98.0+%, TCI America™
SDP

Catalog No. M27895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27895G 5 g
M27891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27895G Supplier TCI America Supplier No. M27895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5784-95-2
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD00443483
InChI Key BHCBPEBRFMLOND-UHFFFAOYSA-N
PubChem CID 231244
IUPAC Name 2-(4-methoxyphenyl)-1H-indole
SMILES COC1=CC=C(C=C1)C2=CC3=CC=CC=C3N2

Specifications

Melting Point 231°C
Quantity 5 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Methoxyphenyl)indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.