Learn More
Sigma Aldrich 2-[(4-Methyl-2-oxo-2H-chromen-7-yl)oxy]ethanethioamide
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- The carbonyl-scavenging ability of 2-amino-1-methylbenzimidazole has been investigated
- Mechanism of menshutkin reaction between 2-amino-1-methylbenzimidazole and iodomethane has been studied in gas phase and in liquid acetonitrile
- It is reported to form adducts with natural allyl, phenethyl, and benzyl isothiocyanates.2-amino-1-methylbenzimidazole may be used for the preparation of 2- or 3-carboxy-4h-pyrimido[2,1-b]-benzazol-4-ones and novel functionalized spiropyranαs derivatives of 2h-1,3-benzoxazinone series
Specifications
Specifications
| Quantity | 5 g |
| Molecular Formula | C12H11NO3S |
| Linear Formula | C12H11NO3S |
| MDL Number | MFCD08443673 |
| Molecular Weight (g/mol) | 249.29 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.