Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-methylphenyl)-1,2-benzisothiazol-3(2H)-one | 2514-30-9 | MFCD05856231 | 99.5% | 241.31 g/mol | C14H11NOS | 25 MG
SDP

Catalog No. 5000246477
Encompass_Preferred

PBIT is a research-grade small-molecule inhibitor of the Jumonji AT-rich interactive domain 1 (JARID1/KDM5) family of histone demethylases, showing activity against JARID1B (IC50 ≈ 3 μM). Supplied as a powder for laboratory use in epigenetics and chromatin biology research.

  • Selective inhibitor of JARID1/KDM5 histone demethylases.
  • Activity against JARID1B with IC50 ≈ 3 μM.
  • High purity (99.5%).
  • Powder form suitable for preparing solutions.
  • Recommended storage: powder -20°C; in solvent -80°C for long-term storage.
  • Includes datasheet with molecular formula and molecular weight.

Catalog No. 50-002-46477 Supplier Medchemexpress LLC Supplier No. HY10145125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PBIT is a research-grade small-molecule inhibitor of the Jumonji AT-rich interactive domain 1 (JARID1/KDM5) family of histone demethylases, showing activity against JARID1B (IC50 ≈ 3 μM). Supplied as a powder for laboratory use in epigenetics and chromatin biology research.

  • Selective inhibitor of JARID1/KDM5 histone demethylases.
  • Activity against JARID1B with IC50 ≈ 3 μM.
  • High purity (99.5%).
  • Powder form suitable for preparing solutions.
  • Recommended storage: powder -20°C; in solvent -80°C for long-term storage.
  • Includes datasheet with molecular formula and molecular weight.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.