Learn More
Medchemexpress LLC 2-(4-methylphenyl)sulfonyl-5-nitrofuran | 343351-67-7 | MFCD01925132 | 99.9% | C11H9NO5S | 10MG
SDP
Catalog No. 5000259867
Encompass_Preferred
NSC697923 is a research-grade small-molecule inhibitor of UBE2N (ubiquitin-conjugating enzyme E2 N, Ubc13) used to block Ubc13-mediated polyubiquitination in biochemical and cellular studies. It is supplied as a white to light-yellow solid with high purity, exhibits good solubility in DMSO, and is stable when stored as a powder at recommended cold temperatures.
  • Selective inhibition of UBE2N (Ubc13) activity in vitro and in cells.
  • High purity suitable for biochemical and cellular assays.
  • White to light-yellow solid with consistent appearance.
  • High solubility in DMSO for preparation of concentrated stock solutions.
  • Formulation guidance provided for in vivo administration.
  • Stable as a powder under recommended cold storage conditions.
Catalog No. 50-002-59867 Supplier Medchemexpress LLC Supplier No. HY1381110MG
NSC697923 is a research-grade small-molecule inhibitor of UBE2N (ubiquitin-conjugating enzyme E2 N, Ubc13) used to block Ubc13-mediated polyubiquitination in biochemical and cellular studies. It is supplied as a white to light-yellow solid with high purity, exhibits good solubility in DMSO, and is stable when stored as a powder at recommended cold temperatures.
  • Selective inhibition of UBE2N (Ubc13) activity in vitro and in cells.
  • High purity suitable for biochemical and cellular assays.
  • White to light-yellow solid with consistent appearance.
  • High solubility in DMSO for preparation of concentrated stock solutions.
  • Formulation guidance provided for in vivo administration.
  • Stable as a powder under recommended cold storage conditions.