Learn More
eMolecules​ 955041-81-3 | [2-(4-Methylphenyl)ethyl]magnesium bromide, 0.5 M in THF | Synthonix - Stock | MFCD07698769 | 223.396 | C9H11BrMg | 0.000 | [Br-].Cc1ccc(CC[Mg+])cc1 | 25ml | 437237313
Catalog No. 502697573 Shop All eMolecules Inc. Products
Encompass_Preferred
[2-(4-Methylphenyl)ethyl]magnesium bromide, 0.5 M in THF | Synthonix - Stock | 955041-81-3 | MFCD07698769 | 223.396 | C9H11BrMg | 0.000 | [Br-].Cc1ccc(CC[Mg+])cc1 | 25ml | 437237313
Catalog No. 50-269-7573 Supplier eMolecules​ Supplier No. 437237313
Add to Cart
Add to Cart
[2-(4-Methylphenyl)ethyl]magnesium bromide, 0.5 M in THF | Synthonix - Stock | 955041-81-3 | MFCD07698769 | 223.396 | C9H11BrMg | 0.000 | [Br-].Cc1ccc(CC[Mg+])cc1 | 25ml | 437237313