Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methylpiperazino)benzaldehyde, ≥95%, Thermo Scientific™

Catalog No. CC36004EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC36004EA 10 g
CC36004CB 250 mg
CC36004DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC36004EA Supplier Thermo Scientific Chemicals Supplier No. CC36004EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85803-62-9
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD01313808
InChI Key GSRYZPWIWYYROI-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-yl benzaldehyde, 2-4-methylpiperazino benzaldehyde, benzaldehyde,2-4-methyl-1-piperazinyl, 4-methylpiperazin-1-ylbenzaldehyde, 2-4-methylpiperazinyl benzaldehyde, 2-4-methylpiperazinyl-benzaldehyde, 2-4-methyl-1-piperazinyl benzaldehyde, 2-4methyl-1-piperazinyl-benzaldehyde, 2-4-methyl-1-piperazinyl-benzaldehyde, 2-4-methylpiperazin-1-yl-benzaldehyde
PubChem CID 2795566
IUPAC Name 2-(4-methylpiperazin-1-yl)benzaldehyde
SMILES CN1CCN(CC1)C2=CC=CC=C2C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 204.27
Percent Purity 95%
Chemical Name or Material 2-(4-Methylpiperazin-1-yl)benzaldehyde
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.