Learn More
USA Scientific Inc 2.4 mm Stainless Steel Beads
Catalog No. 502745184
Encompass
Stainless Steel Beads, 2.4 mm, made of Type 316 stainless steel. For use in bead beating procedures.
Catalog No. 50-274-5184 Supplier USA Scientific Inc Supplier No. 74002702
Stainless Steel Beads, 2.4 mm, made of Type 316 stainless steel. For use in bead beating procedures.