Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Nitrophenoxy)naphthalene 98.0+%, TCI America™
SDP

Catalog No. N08071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08071G Supplier TCI America Supplier No. N08071G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71311-82-5
Molecular Formula C16H11NO3
Molecular Weight (g/mol) 265.268
MDL Number MFCD00427410
InChI Key ORYXOTYJSALBQZ-UHFFFAOYSA-N
PubChem CID 762478
IUPAC Name 2-(4-nitrophenoxy)naphthalene
SMILES C1=CC=C2C=C(C=CC2=C1)OC3=CC=C(C=C3)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 265.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Nitrophenoxy)naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.