Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Nonadecadiynoic Acid 97.0+%, TCI America™
SDP

Catalog No. N0515100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0515100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0515100MG Supplier TCI America Supplier No. N0515100MG

Chemical Identifiers

CAS 76709-55-2
Molecular Formula C19H30O2
Molecular Weight (g/mol) 290.45
MDL Number MFCD00060116
InChI Key LFKNZQNITZRUEE-UHFFFAOYSA-N
PubChem CID 19067421
IUPAC Name nonadeca-2,4-diynoic acid
SMILES CCCCCCCCCCCCCCC#CC#CC(O)=O

Specifications

Melting Point 69°C
Quantity 100 mg
Formula Weight 290.45
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Nonadecadiynoic Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.