Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-nonylphenoxy)acetic acid-d2 | 1219798-75-0 | MFCD00016818 | 280.40 g/mol | C17H24D2O3 | 5 MG
SDP

Catalog No. 5000281903
Encompass_Preferred

2-(4-Nonylphenoxy)acetic acid-d2 is the deuterium-labeled analogue of 2-(4-nonylphenoxy)acetic acid supplied as a solid stable-isotope standard for research use. It is intended for use as an internal standard or tracer in analytical workflows such as mass spectrometry, quantification assays, and method development requiring isotopic differentiation.

  • Deuterium-labeled internal standard for mass spectrometry.
  • Molecular formula C17H24D2O3 and molecular weight 280.40 g/mol for precise mass calculations.
  • Solid powder form suitable for analytical preparation.
  • Available in small weighed quantities for analytical workflows.
  • Recommended storage: powder at -20°C; in solution store at -80°C for long-term stability.

Catalog No. 50-002-81903 Supplier Medchemexpress LLC Supplier No. HYW282887S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(4-Nonylphenoxy)acetic acid-d2 is the deuterium-labeled analogue of 2-(4-nonylphenoxy)acetic acid supplied as a solid stable-isotope standard for research use. It is intended for use as an internal standard or tracer in analytical workflows such as mass spectrometry, quantification assays, and method development requiring isotopic differentiation.

  • Deuterium-labeled internal standard for mass spectrometry.
  • Molecular formula C17H24D2O3 and molecular weight 280.40 g/mol for precise mass calculations.
  • Solid powder form suitable for analytical preparation.
  • Available in small weighed quantities for analytical workflows.
  • Recommended storage: powder at -20°C; in solution store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.