Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-nonylphenoxy)acetic acid-d2 | 1219798-75-0 | MFCD00016818 | 99.0% | 280.40 | C17H24D2O3 | 25 MG
SDP

Catalog No. 5000282086
Encompass_Preferred

2-(4-Nonylphenoxy)acetic acid-d2 is the deuterium-labeled analogue of 2-(4-nonylphenoxy)acetic acid supplied as a solid stable-isotope standard for research and analytical workflows. It is intended for use as an internal standard in mass spectrometry, isotope-tracing, and quantitative studies.

  • Deuterium-labeled analogue for analytical applications.
  • Molecular formula C17H24D2O3; molecular weight 280.40.
  • CAS number 1219798-75-0 for precise identification.
  • Supplied as a 25 mg solid package.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Suitable as an internal standard for mass spectrometry and tracer experiments.

Catalog No. 50-002-82086 Supplier Medchemexpress LLC Supplier No. HYW282887S25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-(4-Nonylphenoxy)acetic acid-d2 is the deuterium-labeled analogue of 2-(4-nonylphenoxy)acetic acid supplied as a solid stable-isotope standard for research and analytical workflows. It is intended for use as an internal standard in mass spectrometry, isotope-tracing, and quantitative studies.

  • Deuterium-labeled analogue for analytical applications.
  • Molecular formula C17H24D2O3; molecular weight 280.40.
  • CAS number 1219798-75-0 for precise identification.
  • Supplied as a 25 mg solid package.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Suitable as an internal standard for mass spectrometry and tracer experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.