Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-nonylphenoxy)acetic acid-d2 | 1219798-75-0 | MFCD03095647 | >98.0% | 280.40 | C17H24D2O3 | 10 MG
SDP

Catalog No. 5000282477
Encompass_Preferred

2-(4-Nonylphenoxy)acetic acid-d2 is the deuterium-labeled analogue of 2-(4-nonylphenoxy)acetic acid supplied for research use. It is intended as an isotopic tracer or internal standard for analytical, metabolic, and pharmacokinetic studies, and is provided as a powder requiring low-temperature storage to preserve isotopic integrity.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Molecular formula C17H24D2O3; molecular weight 280.40.
  • Stable isotope labeling enables mass spectrometric differentiation from unlabeled analytes.
  • Powder form with recommended storage at -20°C for long-term stability.
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-82477 Supplier Medchemexpress LLC Supplier No. HYW282887S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(4-Nonylphenoxy)acetic acid-d2 is the deuterium-labeled analogue of 2-(4-nonylphenoxy)acetic acid supplied for research use. It is intended as an isotopic tracer or internal standard for analytical, metabolic, and pharmacokinetic studies, and is provided as a powder requiring low-temperature storage to preserve isotopic integrity.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Molecular formula C17H24D2O3; molecular weight 280.40.
  • Stable isotope labeling enables mass spectrometric differentiation from unlabeled analytes.
  • Powder form with recommended storage at -20°C for long-term stability.
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.