Learn More
Medchemexpress LLC 2-(4-oxo-1,2,3-benzotriazin-3-yl)-N-[(1S)-1-[4-(trifluoromethoxy)phenyl]ethyl]acetamide | 1929519-13-0 | MFCD32858822 | 99.9% | C18H15F3N4O3 | 10MG
SDP
Catalog No. 5000261392
Encompass_Preferred
Zelatriazin is a potent, selective small-molecule GPR139 agonist supplied for research use. It exhibits reported activity (EC50 ≈ 22 nM) and is provided as a purified compound for in vitro and preclinical pharmacology studies; formulations include solid vials and ready-to-use DMSO solutions.
  • Potent GPR139 agonist (EC50 ≈ 22 nM).
  • High purity (~99.9%).
  • Molecular formula C18H15F3N4O3; mol wt 392.3 g/mol.
  • Available as solid and DMSO solution formats.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-61392 Supplier Medchemexpress LLC Supplier No. HY13222810MG
Zelatriazin is a potent, selective small-molecule GPR139 agonist supplied for research use. It exhibits reported activity (EC50 ≈ 22 nM) and is provided as a purified compound for in vitro and preclinical pharmacology studies; formulations include solid vials and ready-to-use DMSO solutions.
  • Potent GPR139 agonist (EC50 ≈ 22 nM).
  • High purity (~99.9%).
  • Molecular formula C18H15F3N4O3; mol wt 392.3 g/mol.
  • Available as solid and DMSO solution formats.
  • Intended for research use only; not for human or veterinary use.