Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Pentadecadiynoic Acid, TCI America™
SDP

Catalog No. P1032100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1032100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1032100MG Supplier TCI America Supplier No. P1032100MG

Chemical Identifiers

CAS 174063-99-1
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00060115
InChI Key XRCJABKXJJKZPH-UHFFFAOYSA-N
PubChem CID 24206711
IUPAC Name pentadeca-2,4-diynoic acid
SMILES CCCCCCCCCCC#CC#CC(=O)O

Specifications

Melting Point 48°C
Color Blue
Quantity 100 mg
Formula Weight 234.34
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Pentadecadiynoic Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.