Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chem-Impex International, Inc. 2,4-Pentanedione | MFCD00008787 | 1L

Catalog No. 50495362
Encompass_Preferred

2,4-Pentanedione, MFCD00008787, 1L

This item is not returnable. View return policy
Catalog No. 50-495-362 Supplier Chem-Impex International, Inc. Supplier No. 023461L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4-Pentanedione, MFCD00008787, 1L

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.