Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(4-(Piperidin-4-Yl)Phenyl)Acetic Acid Hydrochloride

Catalog No. 917605100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-760-5100MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-760-5100MG Supplier Sigma Aldrich Supplier No. 917605100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(4-(Piperidin-4-yl)phenyl)acetic acid hydrochloride is a 4-aryl piperidine useful as a semi-flexible linker in PROTAC development for targeted protein degradation. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties.Portal: Building PROTAC™ Degraders for Targeted Protein DegradationCrosslinkers for BioconjugationTechnology Spotlight: Degrader Building Blocks for Targeted Protein DegradationPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 384184-43-2
Molecular Formula C55H67BrN4O12
Linear Formula C55H67BrN4O12
Quantity 5 mg
Molecular Weight (g/mol) 1056.04 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.