Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Piperidinedione 98.0+%, TCI America™
SDP

Catalog No. P22961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22961G 1 g
P2296200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22961G Supplier TCI America Supplier No. P22961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50607-30-2
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.116
MDL Number MFCD08704814
InChI Key RDNZDMDLRIQQAX-UHFFFAOYSA-N
Synonym 2,4-piperidinedione, 2,4-piperadinedione, 2,4-dioxopiperidine, 2,4-diketopiperidine, piperidin-2,4-dione, 2,4 dioxopiperdine, 2, 4-dioxopiperidine, pubchem22543, acmc-209ko8
PubChem CID 10887863
IUPAC Name piperidine-2,4-dione
SMILES C1CNC(=O)CC1=O

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 113.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Piperidinedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.