Learn More
Selleck Chemical LLC 2 4-Pyridinedicarboxylic acid
SDP
Supplier:  Selleck Chemical LLC S590625MG
Encompass_Preferred
2 4-Pyridinedicarboxylic acid (Lutidinic Acid 2 4-Dicarboxypyridine 2 4-PDCA) is a compound that structurally mimics 2-oxoglutarate (2-OG) and chelates zinc thus affecting a range of enzymes It blocks the activity of 2-OG oxygenases
Catalog No. 50-000-03750