Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Pyridinedicarboxylic Acid Hydrate, TCI America™
SDP

Catalog No. P055325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P055325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P055325G Supplier TCI America Supplier No. P055325G

Specifications

CAS 499-80-9
Melting Point 243°C
Color White-Yellow
Molecular Formula C7H5NO4
MDL Number MFCD00149413
Quantity 25 g
Synonym 2,4-pyridinedicarboxylic acid, lutidinic acid, 2,4-lutidinic acid, 2,4-pyridinedicarboxylicacid, hydrate 1:1, 4nrq, pd2, 4-carboxypicolinic acid, pubchem16314, pyridine carboxylate, 6a, acmc-1bnb9
InChI Key MJIVRKPEXXHNJT-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=NC(=C1)C(O)=O
IUPAC Name pyridine-2,4-dicarboxylic acid
Molecular Weight (g/mol) 167.12
PubChem CID 10365
ChEBI CHEBI:44737
Formula Weight 167.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Pyridinedicarboxylic Acid Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.