Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Pyridyl)malondialdehyde, 95%

Catalog No. ac425340025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425340025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC425340025 Supplier Thermo Scientific Chemicals Supplier No. 425340025
Only null left
Add to Cart

CAS: 51076-46-1 | C8H7NO2 | 149.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51076-46-1
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00216535
InChI Key RYYVVCNGQOENKM-UHFFFAOYSA-N
Synonym 2-4-pyridyl malondialdehyde, 2-pyridin-4-yl malonaldehyde, 2-pyridin-4-yl propanedial, propanedial, 4-pyridinyl, 2-4-pyridinyl malonaldehyde, 2-pyridin-4-yl malondialdehyde, zlchem 842, 2-4-pyridyl propanedial, ksc925a8h, 2-4-pyridyl propane-1,3-dial
PubChem CID 2737246
IUPAC Name 2-pyridin-4-ylpropanedial
SMILES C1=CN=CC=C1C(C=O)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >320.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min
Packaging Glass bottle
Quantity 2.5 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 149.15
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-(4-Pyridyl)malondialdehyde, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact len

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.