Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-Pyrimidinediamine, N2-[4-[(3-aminopropyl)amino]phenyl]-N4-(5-cyclopropyl-1H-pyrazol-3-yl)- | 2700435-52-3 | 99.5% | 400.91 | 1 ML
SDP

Catalog No. 5000388166
Encompass_Preferred

VEGFR-2-IN-5 hydrochloride is a potent inhibitor of VEGFR2, appearing as an off-white to light yellow solid. It is designed for research applications.

  • Purity: 99.51% (LCMS)
  • Molecular formula: C19H25ClN8
  • Molecular weight: 400.91
  • Store powder at 4°C under nitrogen or at -20°C for up to 3 years.
  • Store in solvent at -80°C for up to 6 months or at -20°C for up to 1 month.
  • Research use only.
  • Referenced in patent WO2013055780A1.

Catalog No. 50-003-88166 Supplier Medchemexpress LLC Supplier No. HY18625A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VEGFR-2-IN-5 hydrochloride is a potent inhibitor of VEGFR2, appearing as an off-white to light yellow solid. It is designed for research applications.

  • Purity: 99.51% (LCMS)
  • Molecular formula: C19H25ClN8
  • Molecular weight: 400.91
  • Store powder at 4°C under nitrogen or at -20°C for up to 3 years.
  • Store in solvent at -80°C for up to 6 months or at -20°C for up to 1 month.
  • Research use only.
  • Referenced in patent WO2013055780A1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.