Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 2-(4-Sulfophenylazo)1,8-dihydroxy-3,6-naphthalenedisulfonic Acid, Trisodium Salt, Indicator Grade

Catalog No. AC151571000
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151571000 100 g Glass bottle
AC151570100 10 g Glass bottle
AC151570250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC151571000 Supplier Thermo Scientific Chemicals Supplier No. 151571000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23647-14-5 | C16H9N2Na3O11S3 | 570.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23647-14-5
Molecular Formula C16H9N2Na3O11S3
Molecular Weight (g/mol) 570.39
MDL Number MFCD00003952
InChI Key LSAWBVPBZOEGOT-UHFFFAOYSA-L
Synonym spadns, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-4-sulfophenyl azo-, trisodium salt, trisodium 4,5-dihydroxy-3-e-2-4-sulfonatophenyl diazen-1-yl naphthalene-2,7-disulfonate, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-2-4-sulfophenyl diazenyl-, sodium salt 1:3, 2-p-sulfophenylazo-1,8-dihydroxy-3,6-naph-thalenedisulfonic acid
PubChem CID 73658865
IUPAC Name trisodium;5-hydroxy-4-oxo-3-[(4-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)O.[Na+].[Na+].[Na+]

Specifications

Color Brown-Red
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 8859
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in organic solvents
Formula Weight 570.39
Grade Indicator
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Sulfophenylazo)1,8-dihydroxy-3,6-naphthalenedisulfonic acid, trisodium salt
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 245-803-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.