Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-tert-Butylbenzyl)propionaldehyde, ≥96.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019956 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9956 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9956 Supplier MilliporeSigma Supelco Supplier No. 4388410MG
Add to Cart
Add to Cart

2-(4-tert-Butylbenzyl)propionaldehyde is categorized under the synthetic fragrance class of compounds widely utilized in consumer products such as perfumes, after shave lotions, cosmetics, etc.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 80-54-6
Density 0.946 g/mL (at 20°C)
Linear Formula C14H20 O
MDL Number MFCD00047655
Synonym 4-tert-Butyl-alpha-methyl-benzenepropanal; 4-tert-Butyl-alpha-methyl-hydrocinnamaldehyde; Butylphenyl methylpropional; Lilial
Type Analytical Standard
Quantity 10 mg
Formula Weight 204.31
Percent Purity ≥96.0% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.