Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(4-trifluoromethylphenyl)piperazine

Catalog No. CDS009461 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS009461 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS009461 Supplier Sigma Aldrich Supplier No. CDS009461100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The mass spectra of 2-phenyl-1-propanol was studied by positive- and negative-ion-fast atom bombardment-mass spectrometric ionization technique.2-Phenyl-1-propanol was used to study the synergism between enzyme catalysis and microwave irradiation.

Specifications

Molecular Formula C11H13F3N2
Linear Formula C11H13F3N2
MDL Number MFCD05864678
Quantity 5 g
Molecular Weight (g/mol) 230.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.