Learn More
Cayman Chemical 2,4-Diacetylphloroglucinol, Purity: ≥98%, CAS: 2161-86-6, 250 mg
SDP
Catalog No. 502973308
Encompass_Preferred
A bacterial metabolite with diverse biological activities; active against B. subtillis, E. faecalis, S. aureus, and S. lividans (MICs = 5, 50, 5, and 5 µg/ml, respectively); active against F. oxysporum, P. debaryanum, and R. solani (MICs = 16, 64, and 32-64 µg/ml, respectively); reduces the growth of corn, flax, tobacco, and tomato seedlings (MICs = 16-32, 32, 8, and 16 µg/ml, respectively); suppresses T. basicola-induced black root rot on tobacco plants; inhibits cell growth in A549, MDA-MB-231, and HeLa cells (IC50s = 52, 94, and 141 nM, respectively)
Catalog No. 50-297-3308 Supplier Cayman Chemical Supplier No. 16345250 MG
A bacterial metabolite with diverse biological activities; active against B. subtillis, E. faecalis, S. aureus, and S. lividans (MICs = 5, 50, 5, and 5 µg/ml, respectively); active against F. oxysporum, P. debaryanum, and R. solani (MICs = 16, 64, and 32-64 µg/ml, respectively); reduces the growth of corn, flax, tobacco, and tomato seedlings (MICs = 16-32, 32, 8, and 16 µg/ml, respectively); suppresses T. basicola-induced black root rot on tobacco plants; inhibits cell growth in A549, MDA-MB-231, and HeLa cells (IC50s = 52, 94, and 141 nM, respectively)