Learn More
Cayman Chemical 2 4Dichlorophenoxy Acetic Aci
SDP
Supplier:  Cayman Chemical 1019050 MG
Encompass_Preferred
A synthetic auxin used as a supplement in plant cell culture media; an active ingredient in herbicides that controls root elongation and cell production by disrupting the actin cytoskeleton
Catalog No. 50-288-8446