Learn More
Ace Glass, Inc. Heating tape, 5/8in, 8ft length, 90w at 115v max, Glas-Col;103A FET0.68
SDP
Catalog No. 50977377
Encompass
5/8 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 8 foot length. Can be operated directly through a temperature control at normal outlet voltage. 90w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.
Catalog No. 50-977-377 Supplier Ace Glass, Inc. Supplier No. 1206308
5/8 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 8 foot length. Can be operated directly through a temperature control at normal outlet voltage. 90w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.