Learn More
GOLANDER LLC 2.4X0.92 SILICONE TUBING, 2.4M
Catalog No. NC2261533
Encompass_Preferred
2.4x0.92 Silicone Tubing, 2.4mm ID, 4.24 mm OD, 0.92 mm wall thickness
This item is not returnable. View return policy
Catalog No. NC2261533 Supplier GOLANDER LLC Supplier No. 2.4X0.92 TUBING_20M
2.4x0.92 Silicone Tubing, 2.4mm ID, 4.24 mm OD, 0.92 mm wall thickness