Learn More
MSC Wera 05074710001 1 Piece, 2-1/2 to 11-1/2 In/Lb, Adjustable Torque Limiting Screwdriver 6" Overall Length, 1/4" Drive, 0.5 In/Lb Graduation
Catalog No. NC1747745
Encompass
Wera 05074710001 1 Piece, 2-1/2 to 11-1/2 In/Lb, Adjustable Torque Limiting Screwdriver 6" Overall Length, 1/4" Drive, 0.5 In/Lb Graduation
Catalog No. NC1747745 Supplier MSC Supplier No. 06150585
Add to Cart
Add to Cart
Wera 05074710001 1 Piece, 2-1/2 to 11-1/2 In/Lb, Adjustable Torque Limiting Screwdriver 6" Overall Length, 1/4" Drive, 0.5 In/Lb Graduation