Learn More
Medchemexpress LLC 2,5(1H,6H)-quinolinedione, 7,8-dihydro- | 15450-69-8 | MFCD00766821 | 99.9% | 163.17 | C9H9NO2 | 5 G
SDP
Catalog No. 5000300879
Encompass_Preferred
7,8-Dihydro-2,5(1H,6H)-quinolinedione (also called compound SF32) is a quinolinedione research chemical reported to have positive inotropic activity. It is provided as a characterized solid for laboratory research and pharmacological assays, with analytical data available for purity and stability to support experimental use.
  • Positive inotropic activity reported.
  • High purity confirmed by HPLC (99.86%).
  • Molecular weight 163.17 and formula C9H9NO2 for accurate dosing.
  • Light brown to light yellow solid appearance.
  • Powder stable at -20°C for long-term storage; solvent stability guidelines provided.
  • Intended for research use; not for human or clinical use.
Catalog No. 50-003-00879 Supplier Medchemexpress LLC Supplier No. HYW0177645G
7,8-Dihydro-2,5(1H,6H)-quinolinedione (also called compound SF32) is a quinolinedione research chemical reported to have positive inotropic activity. It is provided as a characterized solid for laboratory research and pharmacological assays, with analytical data available for purity and stability to support experimental use.
  • Positive inotropic activity reported.
  • High purity confirmed by HPLC (99.86%).
  • Molecular weight 163.17 and formula C9H9NO2 for accurate dosing.
  • Light brown to light yellow solid appearance.
  • Powder stable at -20°C for long-term storage; solvent stability guidelines provided.
  • Intended for research use; not for human or clinical use.