Learn More
Medchemexpress LLC 2-[5-[2-(2,3-dihydro-1H-inden-2-ylamino)pyrimidin-5-yl]-1,3,4-oxadiazol-2-yl]-1-(2,4,6,7-tetrah... | 2156655-68-2 | 98.2% | C22H21N9O2 | 10MG
SDP
Catalog No. 5000260250
Encompass_Preferred
A potent small-molecule inhibitor of autotaxin (ATX) with a reported IC50 of 2.4 nM. Supplied as a light yellow solid with high purity for in vitro research, the compound has molecular formula C22H21N9O2 and a molecular weight of 443.46 g/mol. It is highly soluble in DMSO (≈250 mg/mL) and is intended for enzymatic inhibition and cell-based assay applications.
  • High potency against autotaxin (IC50 = 2.4 nM).
  • High purity suitable for research applications.
  • Good solubility in DMSO for stock solution preparation.
  • Light yellow solid form for easy handling and storage.
  • Suitable for in vitro enzymatic and cell-based assays.
Catalog No. 50-002-60250 Supplier Medchemexpress LLC Supplier No. HY13505310MG
A potent small-molecule inhibitor of autotaxin (ATX) with a reported IC50 of 2.4 nM. Supplied as a light yellow solid with high purity for in vitro research, the compound has molecular formula C22H21N9O2 and a molecular weight of 443.46 g/mol. It is highly soluble in DMSO (≈250 mg/mL) and is intended for enzymatic inhibition and cell-based assay applications.
  • High potency against autotaxin (IC50 = 2.4 nM).
  • High purity suitable for research applications.
  • Good solubility in DMSO for stock solution preparation.
  • Light yellow solid form for easy handling and storage.
  • Suitable for in vitro enzymatic and cell-based assays.