Learn More
Sigma Aldrich 2-(5-(3,4-Dimethylphenyl)-4-oxothieno[2,3-d]pyrimidin-3(4H)-yl)acetic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1,3,5-triisopropylbenzene was employed as swelling agent during the synthesis of mesoporous silicas with two-dimensional hexagonal pores
- It was also employed as micelle expander in the synthesis of highly ordered mesoporous sba-15 silica and magnetic mesoporous silica nanoparticles
Specifications
Specifications
| Molecular Formula | C16H14N2O3S |
| Linear Formula | C16H14N2O3S |
| MDL Number | MFCD02968119 |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 314.36 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.