Learn More
Medchemexpress LLC [2-[[5-[(4-fluorophenyl)carbamoyl]pyrimidin-2-yl]sulfanylmethyl]-4-(trifluoromethoxy)phenyl]boronic acid | 1648843-04-2 | 98.7% | 1 ML
SDP
Catalog No. 5000233653
Encompass_Preferred
SX-682 is a small-molecule allosteric inhibitor of chemokine receptors CXCR1 and CXCR2. It has been investigated preclinically and in clinical trials for reducing tumor myeloid-derived suppressor cell recruitment and enhancing anti-tumor immune responses.
  • Potent CXCR1/2 allosteric inhibitor used in research.
  • Investigated in immuno-oncology and tumor microenvironment studies.
  • Reported to reduce myeloid-derived suppressor cell recruitment.
  • Supplied for laboratory research, often as concentrated solutions or milligram-scale powders.
  • Soluble in DMSO at high concentrations for in vitro use.
Catalog No. 50-002-33653 Supplier Medchemexpress LLC Supplier No. HY11933910MM 1ML
SX-682 is a small-molecule allosteric inhibitor of chemokine receptors CXCR1 and CXCR2. It has been investigated preclinically and in clinical trials for reducing tumor myeloid-derived suppressor cell recruitment and enhancing anti-tumor immune responses.
  • Potent CXCR1/2 allosteric inhibitor used in research.
  • Investigated in immuno-oncology and tumor microenvironment studies.
  • Reported to reduce myeloid-derived suppressor cell recruitment.
  • Supplied for laboratory research, often as concentrated solutions or milligram-scale powders.
  • Soluble in DMSO at high concentrations for in vitro use.