Learn More
Medchemexpress LLC 2-[[5-[(4-fluorophenyl)carbamoyl]pyrimidin-2-yl]sulfanylmethyl]-4-(trifluoromethoxy)phenyl boronic acid | 1648843-04-2 | 99.7% | 10 MG
SDP
Catalog No. 5000234135
Encompass_Preferred
SX-682 is an orally bioavailable small-molecule allosteric inhibitor of chemokine receptors CXCR1 and CXCR2 under clinical investigation for oncology indications; it blocks recruitment of myeloid-derived suppressor cells and can enhance T-cell activity.
  • Orally bioavailable small-molecule CXCR1/2 inhibitor
  • Blocks recruitment of myeloid-derived suppressor cells (MDSCs)
  • Enhances T-cell activation and antitumor immune responses
  • Demonstrated activity in preclinical tumor models and clinical trials
  • Supplied with high purity (typical >99%) and available in small-mass packaging for research
Catalog No. 50-002-34135 Supplier Medchemexpress LLC Supplier No. HY11933910MG
SX-682 is an orally bioavailable small-molecule allosteric inhibitor of chemokine receptors CXCR1 and CXCR2 under clinical investigation for oncology indications; it blocks recruitment of myeloid-derived suppressor cells and can enhance T-cell activity.
  • Orally bioavailable small-molecule CXCR1/2 inhibitor
  • Blocks recruitment of myeloid-derived suppressor cells (MDSCs)
  • Enhances T-cell activation and antitumor immune responses
  • Demonstrated activity in preclinical tumor models and clinical trials
  • Supplied with high purity (typical >99%) and available in small-mass packaging for research