Learn More
Medchemexpress LLC 2-((5-((4-fluorophenyl)carbamoyl)pyrimidinyl)sulfanylmethyl)-4-(trifluoromethoxy)phenyl boronic acid | 1648843-04-2 | 98.7% | 25 MG
SDP
Catalog No. 5000234123
Encompass_Preferred
SX-682 (CAS 1648843-04-2) is an orally bioavailable small-molecule allosteric inhibitor of CXCR1 and CXCR2 investigated for oncology research. It blocks recruitment of immunosuppressive myeloid-derived suppressor cells to the tumor microenvironment and can enhance the activity of checkpoint inhibitors.
  • Oral bioavailability and potent CXCR1/2 antagonism
  • Useful for preclinical and clinical oncology research
  • Blocks myeloid-derived suppressor cell recruitment to tumors
  • Well-characterized chemical identity: CAS 1648843-04-2, C19H14BF4N3O4S
  • Analytical purity typically >98% and supplied as a 25 MG vial
Catalog No. 50-002-34123 Supplier Medchemexpress LLC Supplier No. HY11933925MG
SX-682 (CAS 1648843-04-2) is an orally bioavailable small-molecule allosteric inhibitor of CXCR1 and CXCR2 investigated for oncology research. It blocks recruitment of immunosuppressive myeloid-derived suppressor cells to the tumor microenvironment and can enhance the activity of checkpoint inhibitors.
  • Oral bioavailability and potent CXCR1/2 antagonism
  • Useful for preclinical and clinical oncology research
  • Blocks myeloid-derived suppressor cell recruitment to tumors
  • Well-characterized chemical identity: CAS 1648843-04-2, C19H14BF4N3O4S
  • Analytical purity typically >98% and supplied as a 25 MG vial