Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[5-(5-bromo-1,2-dihydro-2-oxo-3H-indol-3-ylidene)-4-oxo-2-thioxo-3-thiazolidinyl]-butanedioic acid | 301356-95-6 | 98.0% | 457.28 g·mol⁻¹ | C15H9BrN2O6S2 | 2 MG
SDP

Catalog No. 5000214916
Encompass_Preferred

CID5721353 is a small-molecule inhibitor of the BCL6 transcriptional repressor characterized for in vitro research. It shows micromolar activity against BCL6 and is supplied as a solid or as a DMSO solution for laboratory use.

  • Inhibits BCL6 with IC50 212 μM.
  • Binding constant Ki 147 μM.
  • Purity 98.0%.
  • Molecular weight 457.28 g·mol⁻¹.
  • Chemical formula C15H9BrN2O6S2.
  • Available as multiple milligram solid sizes and as 10 mM in DMSO solution.

Catalog No. 50-002-14916 Supplier Medchemexpress LLC Supplier No. HY1005022MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CID5721353 is a small-molecule inhibitor of the BCL6 transcriptional repressor characterized for in vitro research. It shows micromolar activity against BCL6 and is supplied as a solid or as a DMSO solution for laboratory use.

  • Inhibits BCL6 with IC50 212 μM.
  • Binding constant Ki 147 μM.
  • Purity 98.0%.
  • Molecular weight 457.28 g·mol⁻¹.
  • Chemical formula C15H9BrN2O6S2.
  • Available as multiple milligram solid sizes and as 10 mM in DMSO solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.