Learn More
Medchemexpress LLC 2,5,6,7,8-pentadeuterio-3-(trideuteriomethyl)naphthalene-1,4-dione | 478171-80-1 | MFCD01317514 | 99.5% | C11D8O2 | 10MG
SDP
Catalog No. 5000260608
Encompass_Preferred
Vitamin K3-d8 is the deuterium-labeled analogue of menadione used as a stable isotope internal standard and tracer for quantitative analytical methods. Supplied as a high-purity powdered reagent, it is intended for precise quantification in research laboratories and is stable under recommended storage conditions.
  • High isotopic enrichment for reliable internal standardization.
  • Purity 99.5% for accurate quantitation.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Stable as a powder at recommended cold storage temperatures for extended periods.
  • Available in small-mass packages suitable for trace-level analyses.
Catalog No. 50-002-60608 Supplier Medchemexpress LLC Supplier No. HY132389S10MG
Vitamin K3-d8 is the deuterium-labeled analogue of menadione used as a stable isotope internal standard and tracer for quantitative analytical methods. Supplied as a high-purity powdered reagent, it is intended for precise quantification in research laboratories and is stable under recommended storage conditions.
  • High isotopic enrichment for reliable internal standardization.
  • Purity 99.5% for accurate quantitation.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Stable as a powder at recommended cold storage temperatures for extended periods.
  • Available in small-mass packages suitable for trace-level analyses.