Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5,6-Trimethylbenzothiazole, TCI America™
SDP

Catalog No. T173810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T173810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T173810G Supplier TCI America Supplier No. T173810G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5683-41-0
Molecular Formula C10H11NS
Molecular Weight (g/mol) 177.265
MDL Number MFCD00128832
InChI Key ZZLZXZFVVOXVRG-UHFFFAOYSA-N
Synonym 2,5,6-trimethylbenzothiazole, 2,5,6-trimethylbenzo d thiazole, benzothiazole, 2,5,6-trimethyl, pubchem20514, acmc-1b075, benzothiazole,2,5,6-trimethyl, zzlzxzfvvoxvrg-uhfffaoysa, benzothiazole, 2,5,6-trimethyl-6ci,7ci,8ci,9ci
PubChem CID 79765
IUPAC Name 2,5,6-trimethyl-1,3-benzothiazole
SMILES CC1=CC2=C(C=C1C)SC(=N2)C

Specifications

Melting Point 85°C
Boiling Point 143°C
Quantity 10 g
Formula Weight 177.27
Physical Form Crystalline Powder
Chemical Name or Material 2,5,6-Trimethylbenzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.