Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5,6-Trimethylbenzoxazole 98.0+%, TCI America™
SDP

Catalog No. T18455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18455G Supplier TCI America Supplier No. T18455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19219-98-8
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00022849
InChI Key PDVDINPEWGXOHX-UHFFFAOYSA-N
Synonym 2,5,6-trimethylbenzoxazole, benzoxazole, 2,5,6-trimethyl, 2,5,6-trimethylbenzo d oxazole, pubchem8696, 2,6-trimethylbenzoxazole, acmc-1c5qd, 2,5,6-trimethylbenzoxazol, benzoxazole,5,6-trimethyl, 2,5,6-trimethyl-benzoxazole, benzoxazole,2,5,6-trimethyl
PubChem CID 87965
IUPAC Name 2,5,6-trimethyl-1,3-benzoxazole
SMILES CC1=NC2=CC(C)=C(C)C=C2O1

Specifications

Melting Point 93°C
Color White
Quantity 5 g
Formula Weight 161.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5,6-Trimethylbenzoxazole
EINECSNumber (5)-0157
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.