Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,5,6-Trimethylthieno[2,3-d]pyrimidin-4(3H)-one

Catalog No. OTV0009501G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0009501G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0009501G Supplier Sigma Aldrich Supplier No. OTV0009501G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5-Methyl-2-hexanol is a non-crystallizable secondary alcohol and its complex relative permittivity has been measured over wide range of temperatures and pressures.5-Methyl-2-hexanol has been used as internal standard for rapid extraction of aroma compounds from grape brandies and aqueous-alcoholic wood extracts by ultrasound.

Specifications

CAS 18593-45-8
Molecular Formula C9H10N2OS
Linear Formula C9H10N2OS
Quantity 25 g
MDL Number MFCD00457518
Molecular Weight (g/mol) 194.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.